Assay Details
Target Gene Details
Entrez Gene ID: | 64657 |
Gene Name: | mitochondrial ribosomal protein S10 |
Gene Aliases: |
1110038B19Rik, AW743084, Rpms10 |
Location: |
Chr.17:47368887-47381418 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrps10 | NM_001146211.1 | 7 | 655 | NP_001139683.1 |
| NM_001146212.1 | 7 | 658 | NP_001139684.1 | |
| NM_183086.2 | 7 | 748 | NP_898909.2 | |
| AK004151.1 | 7 | 652 | ||
| AK136970.1 | 3 | 780 | BAE23191.1 | |
| BC048912.1 | 7 | 751 | AAH48912.1 | |
| BC096043.1 | 6 | 613 | AAH96043.1 | |
| BC096657.1 | 6 | 563 | AAH96657.1 | |
| DV046794.1 | 7 | 669 |
More Information
Set Membership: |
|

Genomic Map