Assay Details
Target Gene Details
Entrez Gene ID: | 77424 |
Gene Name: | serine protease inhibitor, Kazal type-like |
Gene Aliases: |
9530002K18Rik |
Location: |
Chr.18:44166358-44176707 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spinkl | NM_183123.2 | NP_898946.1 | ||
| AK020535.1 | BAC25634.1 |
More Information
Set Membership: |
|

Genomic Map