Assay Details
Target Gene Details
Entrez Gene ID: | 240055 |
Gene Name: | neuralized homolog 1b (Drosophila) |
Gene Aliases: |
C230078M08Rik, EG240055 |
Location: |
Chr.17:26414965-26446349 on Build GRCm38 |
Assay Gene Location: | Overlaps - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Neurl1b | NM_001081656.2 | NP_001075125.1 | ||
| DQ839448.1 | ||||
| GQ414760.1 |
More Information
Set Membership: |
|

Genomic Map