Assay Details
Target Gene Details
Entrez Gene ID: | 17171 |
Gene Name: | MAS1 oncogene |
Gene Aliases: |
Mas-1, Mgra |
Location: |
Chr.17:12838841-12868145 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mas1 | NM_008552.4 | NP_032578.2 | ||
| AK015036.1 | BAB29684.1 | |||
| AK030261.1 | BAC26865.1 | |||
| AK030265.1 | BAC26868.1 |
More Information
Set Membership: |
|

Genomic Map