Assay Details
Target Gene Details
Entrez Gene ID: | 68842 |
Gene Name: | tubby like protein 4 |
Gene Aliases: |
1110057P05Rik, 2210038L17Rik, Tusp |
Location: |
Chr.17:6106827-6240637 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tulp4 | NM_001103181.1 | NP_001096651.1 | ||
| AK043826.1 | ||||
| AK077171.1 | ||||
| AK084820.1 | ||||
| AK163710.1 | ||||
| BB641651.1 | ||||
| CJ071434.1 |

Genomic Map