Assay Details
Target Gene Details
Entrez Gene ID: | 382044 |
Gene Name: | carboxylesterase 1B |
Gene Aliases: |
Gm5158 |
Location: |
Chr.8:93056725-93080021 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ces1b | NM_001081372.1 | NP_001074841.1 |
More Information
Set Membership: |
|

Genomic Map