Assay Details
Target Gene Details
Entrez Gene ID: | 13884 |
Gene Name: | carboxylesterase 1C |
Gene Aliases: |
Ces-N, Ee-1, Ee1, Es-4, Es-N, Es1, Es4, EsN, PESN |
Location: |
Chr.8:93099015-93131283 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ces1c | NM_007954.4 | NP_031980.2 | ||
| AI527022.1 | ||||
| AK050444.1 | 9 | 1177 | ||
| BC028907.1 | AAH28907.1 | |||
| M57960.1 | AAA63297.1 |
More Information
Set Membership: |
|

Genomic Map