Assay Details
Target Gene Details
Entrez Gene ID: | 666589 |
Gene Name: | predicted gene 8179 |
Gene Aliases: |
EG666589 |
Location: |
Chr.8:32151895-32181847 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm8179 | NR_046039.1 | 1 | 66 | |
| AK143160.1 | 1 | 68 |
Target Gene Details
Entrez Gene ID: | 211323 |
Gene Name: | neuregulin 1 |
Gene Aliases: |
6030402G23Rik, ARIA, D230005F13Rik, GGF, GGFII, HRG, HRGalpha, Hgl, NDF, SMDF |
Location: |
Chr.8:31807452-32890583 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nrg1 |

Genomic Map