Assay Details
Target Gene Details
Entrez Gene ID: | 102182 |
Gene Name: | protein arginine methyltransferase 10 (putative) |
Gene Aliases: |
AI931714, Prmt9 |
Location: |
Chr.8:77549387-77581338 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prmt10 | NM_001081240.3 | NP_001074709.1 | ||
| AK045296.1 | 4 | 2407 | BAC32300.1 | |
| AK045315.1 | BAC32307.1 | |||
| AK154546.1 | BAE32670.1 | |||
| AK161023.1 | ||||
| AK205080.1 | ||||
| AK209748.1 | ||||
| AK217115.1 |
More Information
Set Membership: |
|

Genomic Map