Assay Details
Target Gene Details
Entrez Gene ID: | 20719 |
Gene Name: | serine (or cysteine) peptidase inhibitor, clade B, member 6a |
Gene Aliases: |
4930482L21Rik, D330015H01Rik, PI-6, Serpinb6, Spi3, ovalbumin |
Location: |
Chr.13:33917918-34002794 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Serpinb6a | NM_001164117.1 | 8 | 1389 | NP_001157589.1 |
| NM_001164118.1 | 8 | 1443 | NP_001157590.1 | |
| NM_001243192.1 | 8 | 1293 | NP_001230121.1 | |
| NM_009254.3 | 7 | 1221 | NP_033280.1 | |
| AK154700.1 | 8 | 1285 | BAE32772.1 | |
| AK167295.1 | 7 | 1116 | BAE39400.1 | |
| AK169145.1 | 7 | 1114 | BAE40924.1 | |
| BC006766.1 | 8 | 1302 | AAH06766.1 | |
| BC057950.1 | 7 | 1101 | AAH57950.1 | |
| CT010346.1 | 6 | 1049 | CAJ18554.1 | |
| U25844.1 | 7 | 1116 | AAA79684.1 |
More Information
Set Membership: |
|

Genomic Map