Assay Details
Target Gene Details
Entrez Gene ID: | 622229 |
Gene Name: | solute carrier family 7 (cationic amino acid transporter, y+ system), member pseudogene |
Gene Aliases: |
EG622229 |
Location: |
Chr.3:14363117-14374239 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm6300 | NR_033591.2 | 1 | 233 | |
| AK143764.1 | 1 | 236 |

Genomic Map