Assay Details
Target Gene Details
Entrez Gene ID: | 56185 |
Gene Name: | hydroxyacid oxidase 2 |
Gene Aliases: |
AI325478, Hao-2, Hao3, Haox3 |
Location: |
Chr.3:98874519-98895377 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hao2 | NM_019545.4 | 9 | 1295 | NP_062418.3 |
| AF272947.1 | 9 | 1291 | AAF81795.1 | |
| AK018684.1 | 9 | 1283 | BAB31343.1 | |
| AK078908.1 | 9 | 1285 | BAC37452.1 | |
| AK143991.1 | 9 | 1286 | BAE25651.1 | |
| BC027754.1 | 9 | 1327 | AAH27754.1 |
More Information
Set Membership: |
|

Genomic Map