Assay Details
Target Gene Details
Entrez Gene ID: | 170791 |
Gene Name: | RNA binding motif protein 39 |
Gene Aliases: |
1500012C14Rik, 2310040E03Rik, B330012G18Rik, C79248, R75070, Rnpc2, caper |
Location: |
Chr.2:156147239-156180238 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rbm39 | NM_001291114.1 | 17 | 2385 | NP_001278043.1 |
| NM_001291115.1 | 17 | 2180 | NP_001278044.1 | |
| NM_133242.3 | 17 | 2403 | NP_573505.2 | |
| AK134091.1 | 7 | 955 | BAE22009.1 | |
| AK146046.1 | 17 | 2180 | ||
| AK147076.1 | 17 | 2348 | BAE27657.1 | |
| AK154989.1 | 17 | 2316 | BAE32977.1 | |
| BC004000.1 | 13 | 1735 | AAH04000.1 | |
| BC030493.1 | 15 | 2000 | AAH30493.1 | |
| BC086645.1 | 17 | 2326 | AAH86645.1 | |
| BF465342.1 | 1 | 419 |
More Information
Set Membership: |
|

Genomic Map