Assay Details
Target Gene Details
Entrez Gene ID: | 12623 |
Gene Name: | carboxylesterase 1G |
Gene Aliases: |
Ces-1, Ces1, Ses-1 |
Location: |
Chr.8:93302369-93337209 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ces1g | NM_021456.4 | 14 | 1756 | NP_067431.2 |
| AK136446.1 | 14 | 1753 | BAE22982.1 | |
| AK136588.1 | 14 | 1753 | BAE23063.1 | |
| AK149446.1 | 14 | 1753 | BAE28882.1 | |
| AK162328.1 | 14 | 1731 | BAE36856.1 | |
| BC021150.1 | 14 | 1775 | AAH21150.1 | |
| BC026897.1 | 14 | 1736 | AAH26897.1 | |
| Y12887.1 | 14 | 1742 | CAA73388.1 |
More Information
Set Membership: |
|

Genomic Map