Assay Details
Target Gene Details
Entrez Gene ID: | 100040970 |
Gene Name: | ribosomal protein L14, pseudogene 1 |
Gene Aliases: |
ENSMUSG00000046721, Gm9811 |
Location: |
Chr.7:45324958-45325651 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rpl14-ps1 | NR_110499.1 | 1 | 414 | |
| BC008177.1 | 1 | 278 | AAH08177.1 | |
| BC092249.1 | 1 | 414 | AAH92249.1 |
Target Gene Details
Entrez Gene ID: | 68667 |
Gene Name: | transient receptor potential cation channel, subfamily M, member 4 |
Gene Aliases: |
1110030C19Rik, AW047689, LTRPC4, LTrpC-4, TRPM4B |
Location: |
Chr.7:45303155-45334070 on Build GRCm38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trpm4 | NM_175130.4 | NP_780339.2 | ||
| AJ575814.1 | CAE05940.1 | |||
| BC096475.1 | AAH96475.1 |
More Information
Set Membership: |
|

Genomic Map