Assay Details
Target Gene Details
Entrez Gene ID: | 104443 |
Gene Name: | neuropeptide FF receptor 2 |
Gene Aliases: |
Gpr74, HG31, NPFF2 |
Location: |
Chr.5:89526577-89583914 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Npffr2 | NM_133192.3 | NP_573455.2 | ||
| AF236084.1 | AAK58514.1 | |||
| AF330054.1 | AAK94198.1 | |||
| AK050939.1 | BAC34468.1 |
More Information
Set Membership: |
|

Genomic Map