Assay Details
Target Gene Details
Entrez Gene ID: | 28071 |
Gene Name: | twist basic helix-loop-helix transcription factor 1 neighbor |
Gene Aliases: |
2410173G11, 2810024J17Rik, D16Wsu83e |
Location: |
Chr.12:33429624-33439380 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Twistnb | NM_172253.2 | NP_758457.1 | ||
| AK008677.1 | ||||
| AK010829.1 | BAB27210.1 | |||
| AK016638.1 | ||||
| AK017016.1 | BAC25502.1 | |||
| AK017810.1 | BAB30947.1 | |||
| AK160682.1 | ||||
| AK179512.1 | ||||
| BC139162.1 | ||||
| BC139163.1 |
More Information
Set Membership: |
|

Genomic Map