Assay Details
Target Gene Details
Entrez Gene ID: | 320749 |
Gene Name: | RIKEN cDNA D630041G03 gene |
Gene Aliases: |
- |
Location: |
Chr.7:4467245-4470884 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| D630041G03Rik | NR_028416.1 | 1 | 1377 | |
| AK052741.1 | 1 | 1377 | ||
| AK085348.1 | 1 | 1377 |
Target Gene Details
Entrez Gene ID: | 67425 |
Gene Name: | EPS8-like 1 |
Gene Aliases: |
2310051G19Rik, 4632407K17Rik, AW060268, DRC3, EPS8R1 |
Location: |
Chr.7:4460631-4480487 on Build GRCm38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eps8l1 | NM_001290416.1 | NP_001277345.1 | ||
| NM_026146.4 | NP_080422.1 | |||
| AK014556.1 | ||||
| AK019490.2 | BAB31756.2 | |||
| AY074931.1 | AAL76120.1 |
More Information
Set Membership: |
|

Genomic Map