Assay Details
Target Gene Details
Entrez Gene ID: | 16625 |
Gene Name: | serine (or cysteine) peptidase inhibitor, clade A, member 3C |
Gene Aliases: |
1A1, Kalbp, Klkbp, spi2 |
Location: |
Chr.12:104146907-104153933 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Serpina3c | NM_008458.2 | NP_032484.1 | ||
| BC116724.1 |
More Information
Set Membership: |
|

Genomic Map