Assay Details
Target Gene Details
Entrez Gene ID: | 56745 |
Gene Name: | C1q and tumor necrosis factor related protein 1 |
Gene Aliases: |
1600017K21Rik, CTRP1, Zsig37 |
Location: |
Chr.11:118428457-118451782 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C1qtnf1 | NM_001204129.1 | NP_001191058.1 | ||
| NM_001204130.1 | NP_001191059.1 | |||
| NM_019959.3 | NP_064343.1 | |||
| AF192499.1 | AAF06664.1 | |||
| AK005484.1 | BAB24070.1 | |||
| AK145104.1 | BAE26237.1 | |||
| BC049894.1 | AAH49894.1 | |||
| CJ056741.1 | ||||
| DQ002394.1 | AAY21926.1 |
Target Gene Details
Entrez Gene ID: | 100126224 |
Gene Name: | predicted gene 11747 |
Gene Aliases: |
OTTMUSG00000003972 |
Location: |
Chr.11:118444200-118454995 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm11747 | NR_045902.1 | |||
| AK033110.1 |
More Information
Set Membership: |
|

Genomic Map