Assay Details
Target Gene Details
Entrez Gene ID: | 104382 |
Gene Name: | BarH-like 2 (Drosophila) |
Gene Aliases: |
E130309B19Rik, MBH1 |
Location: |
Chr.5:106452523-106458166 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Barhl2 | NM_001005477.1 | NP_001005477.1 | ||
| AK053799.1 | ||||
| BC055789.1 | AAH55789.1 | |||
| BC078444.1 | AAH78444.1 |
More Information
Set Membership: |
|

Genomic Map