Assay Details
Target Gene Details
Entrez Gene ID: | 11732 |
Gene Name: | progressive ankylosis |
Gene Aliases: |
Ankh, D15Ertd221e, ank, mKIAA1581 |
Location: |
Chr.15:27466677-27594909 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ank | NM_020332.4 | NP_065065.3 | ||
| AF001532.1 | AAB65653.1 | |||
| AF274752.1 | AAF88038.1 | |||
| AK083135.1 | BAC38778.1 | |||
| AK148070.1 | BAE28324.1 | |||
| BC054379.1 | AAH54379.1 | |||
| DQ832285.1 |
More Information
Set Membership: |
|

Genomic Map