Assay Details
Target Gene Details
Entrez Gene ID: | 70920 |
Gene Name: | RIKEN cDNA 4921511H03 gene |
Gene Aliases: |
- |
Location: |
Chr.5:7302505-7311491 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4921511H03Rik | NM_027603.2 | 2 | 412 | NP_081879.1 |
| AK014870.1 | 2 | 729 | BAB29593.1 | |
| AK076678.1 | 2 | 413 | BAC36441.1 | |
| AK133311.1 | 2 | 445 | BAE21604.1 | |
| BC049718.1 | 2 | 449 | AAH49718.1 |
Target Gene Details
Entrez Gene ID: | 207618 |
Gene Name: | zinc finger protein 804B |
Gene Aliases: |
Gm441 |
Location: |
Chr.5:6769030-7344378 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp804b | NM_001163223.1 | NP_001156695.1 |
More Information
Set Membership: |
|

Genomic Map