Assay Details
Target Gene Details
Entrez Gene ID: | 69672 |
Gene Name: | thioredoxin domain containing 15 |
Gene Aliases: |
2310047H23Rik, AI854086, C5orf14, UNQ335 |
Location: |
Chr.13:55714650-55726227 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Txndc15 | NM_175150.3 | NP_780359.2 | ||
| AK009878.1 | ||||
| AK046071.1 | 1 | 631 | ||
| AK079517.1 | BAC37669.1 | |||
| AK160051.1 | BAE35591.1 | |||
| AK167701.1 | BAE39745.1 | |||
| AK167838.1 | BAE39859.1 | |||
| AK167977.1 | BAE39969.1 | |||
| BC029015.1 | AAH29015.1 | |||
| BC062180.1 | AAH62180.1 | |||
| BC094275.1 | AAH94275.1 |
More Information
Set Membership: |
|

Genomic Map