Assay Details
Target Gene Details
Entrez Gene ID: | 97895 |
Gene Name: | NLR family, pyrin domain containing 4F |
Gene Aliases: |
C330026N02Rik, C86045, Nalp-kappa, Nalp4f |
Location: |
Chr.13:65177111-65205716 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nlrp4f | NM_175290.4 | NP_780499.3 | ||
| AK049352.1 | BAC33701.1 | |||
| AY596201.1 | AAU06322.1 | |||
| BC098479.1 | AAH98479.1 |
More Information
Set Membership: |
|

Genomic Map