Assay Details
Target Gene Details
Entrez Gene ID: | 268288 |
Gene Name: | sterile alpha motif domain containing 3 |
Gene Aliases: |
Gm623 |
Location: |
Chr.10:26229618-26272916 on Build GRCm38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Samd3 | NM_001013766.2 | NP_001013788.2 | ||
| NM_001115154.1 | NP_001108626.1 | |||
| AK082201.1 | BAC38435.1 | |||
| AK144462.1 | ||||
| BC150876.1 |
More Information
Set Membership: |
|

Genomic Map