Assay Details
Target Gene Details
Entrez Gene ID: | 68828 |
Gene Name: | syncoilin |
Gene Aliases: |
1110057H03Rik, SNIP4 |
Location: |
Chr.4:129287536-129309386 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sync | NM_023485.3 | NP_075974.3 | ||
| AJ251641.1 | CAC17787.1 | |||
| AK081983.1 | BAC38387.1 | |||
| AK160756.1 | BAE35992.1 | |||
| BC106855.2 | AAI06856.1 | |||
| BY728583.1 | ||||
| FJ752162.1 | ||||
| FJ752163.1 | ||||
| FJ752164.1 |
More Information
Set Membership: |
|

Genomic Map