Assay Details
Target Gene Details
Entrez Gene ID: | 22344 |
Gene Name: | vascular endothelial zinc finger 1 |
Gene Aliases: |
AI848691, AW046909, db1 |
Location: |
Chr.11:88068256-88084729 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vezf1 | NM_016686.4 | NP_057895.2 | ||
| AF104410.3 | AAD02883.1 |
More Information
Set Membership: |
|

Genomic Map