Assay Details
Target Gene Details
Entrez Gene ID: | 66761 |
Gene Name: | RIKEN cDNA 4933417A18 gene |
Gene Aliases: |
- |
Location: |
Chr.13:34924405-34956218 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4933417A18Rik | NM_025750.3 | NP_080026.2 | ||
| AK016778.1 | BAB30424.1 | |||
| AK016838.1 | BAB30456.1 | |||
| BC100450.1 | AAI00451.1 | |||
| BC145747.1 |
More Information
Set Membership: |
|

Genomic Map