Assay Details
Target Gene Details
Entrez Gene ID: | 78885 |
Gene Name: | coronin 7 |
Gene Aliases: |
0610011B16Rik, AW048373, AW556392 |
Location: |
Chr.16:4626884-4679720 on Build GRCm38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Coro7 | NM_030205.4 | NP_084481.3 | ||
| AK018739.1 | BAB31380.1 | |||
| AK088155.1 | ||||
| AK143624.1 | ||||
| BC061006.1 | AAH61006.1 |
Target Gene Details
Entrez Gene ID: | 246154 |
Gene Name: | vasorin |
Gene Aliases: |
2610528G05Rik, Atia, Slitl2 |
Location: |
Chr.16:4639945-4651166 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vasn | NM_139307.3 | NP_647468.2 | ||
| AJ458938.1 | CAD30331.1 | |||
| AK012169.1 | BAB28075.1 | |||
| AK083684.1 | BAC38992.1 | |||
| AK132325.1 | BAE21105.1 | |||
| BC050274.1 | AAH50274.1 |
More Information
Set Membership: |
|

Genomic Map