Assay Details
Target Gene Details
Entrez Gene ID: | 20091 |
Gene Name: | ribosomal protein S3A1 |
Gene Aliases: |
Rps3a |
Location: |
Chr.3:86137940-86142668 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rps3a1 | NM_016959.4 | NP_058655.3 | ||
| AK003158.1 | BAB22611.1 | |||
| AK010605.1 | BAB27055.1 | |||
| AK012345.1 | BAB28176.1 | |||
| AK050610.1 | BAC34341.1 | |||
| AK088113.1 | BAC40152.1 | |||
| AK135318.1 | BAE22486.1 | |||
| AK146304.1 | BAE27060.1 | |||
| AK148513.1 | BAE28594.1 | |||
| AK150441.1 | BAE29563.1 | |||
| AK150796.1 | BAE29859.1 | |||
| AK150814.1 | BAE29877.1 | |||
| AK151429.1 | BAE30393.1 | |||
| AK151982.1 | BAE30849.1 | |||
| AK153480.1 | BAE32029.1 | |||
| AK159777.1 | BAE35361.1 | |||
| AK166713.1 | ||||
| AK167512.1 | BAE39587.1 | |||
| AK167562.1 | BAE39625.1 | |||
| AK168240.1 | BAE40190.1 | |||
| AK190515.1 | ||||
| AK196258.1 | ||||
| AK200497.1 | ||||
| AK207259.1 | ||||
| AK207595.1 | ||||
| AK207749.1 | ||||
| AK207971.1 | ||||
| AK208520.1 | ||||
| AK208628.1 | ||||
| AL118161.1 | ||||
| BC039659.1 | AAH39659.1 | |||
| BC081451.1 | AAH81451.1 | |||
| BC083338.1 | AAH83338.1 | |||
| BC084675.1 | AAH84675.1 | |||
| BC106115.1 | AAI06116.1 | |||
| M88335.1 |
More Information
Set Membership: |
|

Genomic Map