Assay Details
Target Gene Details
Entrez Gene ID: | 57815 |
Gene Name: | spermatogenesis associated 5 |
Gene Aliases: |
2510048F20Rik, C78064, Spaf |
Location: |
Chr.3:37420280-37579096 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spata5 | NM_001163511.1 | NP_001156983.1 | ||
| NM_021343.2 | NP_067318.2 | |||
| AF049099.1 | AAD02481.1 | |||
| AK011111.2 | BAB27406.1 | |||
| AK144998.1 | BAE26178.1 | |||
| BC145302.1 |
More Information
Set Membership: |
|

Genomic Map