Assay Details
Target Gene Details
Entrez Gene ID: | 58875 |
Gene Name: | 3-hydroxyisobutyrate dehydrogenase |
Gene Aliases: |
6430402H10Rik, AI265272 |
Location: |
Chr.6:52546230-52640300 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hibadh | NM_145567.1 | NP_663542.1 | ||
| AB051067.1 | ||||
| AK078175.1 | BAC37162.1 | |||
| BC003914.1 | AAH03914.1 |
More Information
Set Membership: |
|

Genomic Map