Assay Details
Target Gene Details
Entrez Gene ID: | 107971 |
Gene Name: | fibroblast growth factor receptor substrate 3 |
Gene Aliases: |
4930417B13Rik, AI449674, Frs2beta, Snt2 |
Location: |
Chr.17:47687296-47704286 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Frs3 |
Target Gene Details
Entrez Gene ID: | 66119 |
Gene Name: | translocase of outer mitochondrial membrane 6 homolog (yeast) |
Gene Aliases: |
1110002E23Rik, AI663970 |
Location: |
Chr.17:47686645-47688386 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tomm6 | NM_001164729.1 | NP_001158201.1 | ||
| NM_025365.3 | NP_079641.1 | |||
| AK003291.1 | BAB22694.1 | |||
| AK010470.1 | BAB26963.1 | |||
| AK019199.1 | BAB31597.1 | |||
| BC021590.1 | AAH21590.1 | |||
| BC037589.1 | AAH37589.1 | |||
| CA319049.1 | ||||
| CA321392.1 |
More Information
Set Membership: |
|

Genomic Map