Assay Details
Target Gene Details
Entrez Gene ID: | 72254 |
Gene Name: | RIKEN cDNA 1700030K09 gene |
Gene Aliases: |
- |
Location: |
Chr.8:72443880-72460541 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1700030K09Rik | NM_028170.2 | NP_082446.2 | ||
| AK089918.1 | ||||
| AK133815.1 | BAE21858.1 | |||
| AK157359.1 | ||||
| AK172202.1 | ||||
| BC008558.1 | AAH08558.1 |
More Information
Set Membership: |
|

Genomic Map