Assay Details
Target Gene Details
Entrez Gene ID: | 67486 |
Gene Name: | polymerase (RNA) III (DNA directed) polypeptide G |
Gene Aliases: |
2310047G20Rik, AV275904, RPC32, RPC7 |
Location: |
Chr.13:81673837-81711013 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Polr3g | NM_001081176.1 | NP_001074645.1 | ||
| AK009045.1 | ||||
| AK009876.1 | ||||
| AK010126.1 | BAB26717.3 | |||
| AK166165.1 | 3 | 4329 | ||
| BC030063.1 | AAH30063.1 | |||
| BC066818.1 | AAH66818.1 |
More Information
Set Membership: |
|

Genomic Map