Assay Details
Target Gene Details
Entrez Gene ID: | 78239 |
Gene Name: | serine protease inhibitor, Kunitz type 4 |
Gene Aliases: |
9230105I15Rik |
Location: |
Chr.2:164698501-164702448 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spint4 | NM_030058.2 | NP_084334.1 | ||
| AK020315.1 | BAB32069.1 | |||
| AY226988.1 | AAP43945.1 | |||
| BC119299.1 | ||||
| BC119301.1 |
More Information
Set Membership: |
|

Genomic Map