Assay Details
Target Gene Details
Entrez Gene ID: | 434147 |
Gene Name: | RIKEN cDNA D930028M14 gene |
Gene Aliases: |
EG434147 |
Location: |
Chr.7:25152457-25156630 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| D930028M14Rik | NR_045847.1 | |||
| AK142847.1 | BAE25208.1 |
Target Gene Details
Entrez Gene ID: | 18987 |
Gene Name: | POU domain, class 2, transcription factor 2 |
Gene Aliases: |
Oct-2, Oct2a, Oct2b, Otf-2, Otf2 |
Location: |
Chr.7:25091114-25179992 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pou2f2 |
More Information
Set Membership: |
|

Genomic Map