Assay Details
Target Gene Details
Entrez Gene ID: | 68348 |
Gene Name: | serine (or cysteine) peptidase inhibitor, clade A, member 1F |
Gene Aliases: |
0610012A11Rik, AI327419, Serpina2, epserin |
Location: |
Chr.12:103688044-103695529 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Serpina1f | NM_001164742.1 | NP_001158214.1 | ||
| NM_026687.2 | NP_080963.2 | |||
| AB233454.1 | BAE45638.1 | |||
| AK002574.1 | BAB22199.1 | |||
| AK165409.1 | BAE38167.1 | |||
| BC119526.1 |
More Information
Set Membership: |
|

Genomic Map