Assay Details
Target Gene Details
Entrez Gene ID: | 17389 |
Gene Name: | matrix metallopeptidase 16 |
Gene Aliases: |
MT3-MMP, Mt3mmp |
Location: |
Chr.4:17853482-18118734 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mmp16 | NM_019724.3 | NP_062698.2 | ||
| AB021228.1 | BAA78420.1 | |||
| AF282844.1 | AAG17704.1 | |||
| AK046170.1 | ||||
| AK048543.1 | ||||
| AK053598.1 | ||||
| BC057926.1 | AAH57926.1 |
More Information
Set Membership: |
|

Genomic Map