Assay Details
Target Gene Details
Entrez Gene ID: | 30944 |
Gene Name: | zinc finger protein 354C |
Gene Aliases: |
5330421P20Rik, AJ18, AW743325, Kid3, Tcf17l2, Znf354c, mKIAA4218 |
Location: |
Chr.11:50811081-50827731 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp354c | NM_013922.4 | NP_038950.3 | ||
| AF192804.1 | AAF35173.1 | |||
| AK086760.1 | BAC39738.1 | |||
| AK145882.1 | BAE26722.1 | |||
| AK220194.1 | BAD90379.1 | |||
| BC079908.1 | AAH79908.1 |
More Information
Set Membership: |
|

Genomic Map