Assay Details
Target Gene Details
Entrez Gene ID: | 70620 |
Gene Name: | ubiquitin-conjugating enzyme E2 variant 2 |
Gene Aliases: |
1110021H13Rik, 4632410D19Rik, 5730524P06Rik, 6820402M05, AI848315, C81524, MMS2 |
Location: |
Chr.16:15550986-15594518 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ube2v2 | NM_001159351.1 | NP_001152823.1 | ||
| NM_023585.4 | NP_076074.2 | |||
| AF303828.1 | AAG22084.1 | |||
| AK017786.1 | BAB30932.1 | |||
| AK019486.1 | BAB31753.1 | |||
| AK028472.1 | BAC25968.1 | |||
| AK028742.1 | BAC26094.1 | |||
| AK031233.1 | BAC27311.1 | |||
| AK033016.1 | BAC28128.1 | |||
| AK146073.1 | BAE26878.1 | |||
| BC029742.1 | AAH29742.1 | |||
| BC058374.1 | AAH58374.1 | |||
| BC083098.1 | AAH83098.1 | |||
| BC094903.1 | AAH94903.1 | |||
| BY025619.1 | ||||
| CJ049747.1 |
More Information
Set Membership: |
|

Genomic Map