Assay Details
Target Gene Details
Entrez Gene ID: | 66313 |
Gene Name: | SMAD specific E3 ubiquitin protein ligase 2 |
Gene Aliases: |
2810411E22Rik, AI558114, AI649275 |
Location: |
Chr.11:106820064-106920715 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smurf2 | NM_025481.2 | NP_079757.2 | ||
| AK012060.1 | ||||
| AY685230.1 | AAV87906.1 | |||
| BC138786.1 | ||||
| BC138788.1 | ||||
| BU058366.1 |
More Information
Set Membership: |
|

Genomic Map