Assay Details
Target Gene Details
Entrez Gene ID: | 77582 |
Gene Name: | membrane bound O-acyltransferase domain containing 7 |
Gene Aliases: |
5730589L02Rik, BB1, Leng4, Lpiat, Lpiat1, mBB1 |
Location: |
Chr.7:3677789-3693525 on Build GRCm38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mboat7 | NM_029934.3 | NP_084210.2 | ||
| AB055410.1 | BAC53808.1 | |||
| AK019981.1 | BAB31950.1 | |||
| AK038998.1 | ||||
| AK044149.1 | ||||
| AK150010.1 | BAE29235.1 | |||
| AK154606.1 | BAE32708.1 | |||
| AK158998.1 | ||||
| AK170124.1 | BAE41580.1 | |||
| BC023417.1 | AAH23417.1 | |||
| BC118958.2 | ||||
| EU016380.1 |
More Information
Set Membership: |
|

Genomic Map