Assay Details
Target Gene Details
Entrez Gene ID: | 23805 |
Gene Name: | adenomatosis polyposis coli 2 |
Gene Aliases: |
AI852447, APCL, R75424 |
Location: |
Chr.10:80295957-80318257 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Apc2 | NM_011789.2 | NP_035919.2 |
More Information
Set Membership: |
|

Genomic Map