Assay Details
Target Gene Details
Entrez Gene ID: | 19227 |
Gene Name: | parathyroid hormone-like peptide |
Gene Aliases: |
PTH-like, Pthrp |
Location: |
Chr.6:147252110-147264085 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pthlh | NM_008970.3 | NP_032996.2 | ||
| AJ278119.1 | CAC39218.1 | |||
| AY183377.1 | AAO25537.1 | |||
| AY220497.1 | AAO64343.1 | |||
| BC058187.1 | AAH58187.1 | |||
| CB194603.1 |
More Information
Set Membership: |
|

Genomic Map