Assay Details
Target Gene Details
Entrez Gene ID: | 66612 |
Gene Name: | ORM1-like 3 (S. cerevisiae) |
Gene Aliases: |
2810011N17Rik, AI315223 |
Location: |
Chr.11:98581294-98587245 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ormdl3 | NM_025661.4 | NP_079937.1 | ||
| AK009621.1 | BAB26397.1 | |||
| AK012718.1 | BAB28432.1 | |||
| AK087497.1 | BAC39898.1 | |||
| AK154085.1 | BAE32363.1 | |||
| BC046594.1 | AAH46594.1 |
More Information
Set Membership: |
|

Genomic Map