Assay Details
Target Gene Details
Entrez Gene ID: | 73174 |
Gene Name: | TBK1 binding protein 1 |
Gene Aliases: |
3110043L15Rik, ProSAPiP2, SINTBAD |
Location: |
Chr.11:97136171-97151490 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tbkbp1 | NM_198100.2 | NP_932768.2 | ||
| AK014173.1 | ||||
| BC057590.1 | AAH57590.1 |
More Information
Set Membership: |
|

Genomic Map