Assay Details
Target Gene Details
Entrez Gene ID: | 14165 |
Gene Name: | fibroblast growth factor 10 |
Gene Aliases: |
AEY17, BB213776, Fgf-10, Gsfaey17 |
Location: |
Chr.13:118714699-118792573 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fgf10 | NM_008002.4 | NP_032028.1 | ||
| AK034742.1 | BAC28816.1 | |||
| AK045267.1 | BAC32287.1 | |||
| AK085244.1 | BAC39398.1 | |||
| AK141633.1 | BAE24777.1 | |||
| BC048229.1 | AAH48229.1 | |||
| D89080.1 | BAA22836.1 | |||
| U94517.1 | AAD00761.1 |
More Information
Set Membership: |
|

Genomic Map