Assay Details
Target Gene Details
Entrez Gene ID: | 67645 |
Gene Name: | armadillo repeat containing 12 |
Gene Aliases: |
4930511I11Rik, AV041658 |
Location: |
Chr.17:28527534-28539110 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Armc12 | NM_026290.3 | NP_080566.2 | ||
| AK005672.1 | BAB24180.1 | |||
| AK015756.1 | BAB29960.1 | |||
| BC049559.1 | AAH49559.1 | |||
| EU477370.1 |
More Information
Set Membership: |
|

Genomic Map